Learn More
Medchemexpress LLC Adenosine dialdehyde | 34240-05-6 | MFCD00056960 | 99.0% | 265.23 g/mol | C10H11N5O4 | 10MG
SDP
Catalog No. 5000238686
Encompass_Preferred
Adenosine dialdehyde is a purine nucleoside analogue that acts as a potent inhibitor of S-adenosylhomocysteine hydrolase (SAHH). It is used as a biochemical research reagent to investigate methylation pathways and methyltransferase activity, and is supplied in solid milligram quantities and as a 10 mM solution in DMSO for convenient assay preparation.
  • Potent SAHH inhibitor for methylation pathway studies.
  • Supplied in both solid and ready-to-use solution formats.
  • High stated purity for analytical and research applications.
  • Compatible with enzyme inhibition assays and cellular studies.
  • Provided with datasheet, certificate of analysis, and safety data sheet for compliance.
Catalog No. 50-002-38686 Supplier Medchemexpress LLC Supplier No. HY12305510MG
Adenosine dialdehyde is a purine nucleoside analogue that acts as a potent inhibitor of S-adenosylhomocysteine hydrolase (SAHH). It is used as a biochemical research reagent to investigate methylation pathways and methyltransferase activity, and is supplied in solid milligram quantities and as a 10 mM solution in DMSO for convenient assay preparation.
  • Potent SAHH inhibitor for methylation pathway studies.
  • Supplied in both solid and ready-to-use solution formats.
  • High stated purity for analytical and research applications.
  • Compatible with enzyme inhibition assays and cellular studies.
  • Provided with datasheet, certificate of analysis, and safety data sheet for compliance.