Learn More
Medchemexpress LLC Adenosine dialdehyde | 34240-05-6 | MFCD00056960 | 99.0% | 265.23 g/mol | C10H11N5O4 | 10MM 1ML
SDP
Catalog No. 5000238687
Encompass_Preferred
Adenosine dialdehyde is a purine nucleoside analogue that potently inhibits S-adenosylhomocysteine hydrolase (SAHH). It is used in biochemical assays and cancer research to study methylation-dependent pathways and can be supplied as a 10 mM solution in DMSO or as solid analytical standards.
Catalog No. 50-002-38687 Supplier Medchemexpress LLC Supplier No. HY12305510MM 1ML
Adenosine dialdehyde is a purine nucleoside analogue that potently inhibits S-adenosylhomocysteine hydrolase (SAHH). It is used in biochemical assays and cancer research to study methylation-dependent pathways and can be supplied as a 10 mM solution in DMSO or as solid analytical standards.