Learn More
Medchemexpress LLC Adenosine dialdehyde | 34240-05-6 | MFCD00056960 | >98.0% | 265.23 g/mol | C10H11N5O4 | 5MG
SDP
Catalog No. 5000238794
Encompass_Preferred
Adenosine dialdehyde is a purine nucleoside analogue that potently inhibits S-adenosylhomocysteine hydrolase (SAHH). It is used as a research reagent in biochemical and cellular studies to probe methylation pathways and methyltransferase activity. The compound is a crystalline solid with molecular weight 265.23 g/mol and formula C10H11N5O4, and it is soluble in DMSO.
Catalog No. 50-002-38794 Supplier Medchemexpress LLC Supplier No. HY1230555MG
Adenosine dialdehyde is a purine nucleoside analogue that potently inhibits S-adenosylhomocysteine hydrolase (SAHH). It is used as a research reagent in biochemical and cellular studies to probe methylation pathways and methyltransferase activity. The compound is a crystalline solid with molecular weight 265.23 g/mol and formula C10H11N5O4, and it is soluble in DMSO.