Learn More
TARGETMOL CHEMICALS INC ADENOSINE DIALDEHYDE ADO 50MG
SDP
Catalog No. 502580969
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Adenosine Dialdehyde (Periodate-oxidized adenosine) is a purine nucleoside analogue. Adenosine Dialdehyde is a potent inhibitor of S-Adenosylhomocysteine hydrolase (SAHH) (Ki=3.3 nM). Adenosine Dialdehyde exhibits potent anti-tumor activity in vivo. purity: 95%
Catalog No. 50-258-0969 Supplier TARGETMOL CHEMICALS INC Supplier No. T2223150MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Adenosine Dialdehyde (Periodate-oxidized adenosine) is a purine nucleoside analogue. Adenosine Dialdehyde is a potent inhibitor of S-Adenosylhomocysteine hydrolase (SAHH) (Ki=3.3 nM). Adenosine Dialdehyde exhibits potent anti-tumor activity in vivo. purity: 95%