Learn More
Apexbio Technology LLC Adox, 25mg. Cas: 34240-05-6 MFCD: MFCD00063602. indirect methyltransferase inhibitor
SDP
Catalog No. NC1672427
Encompass_Preferred
Adenosine dialdehyde (AdOx) has been identified as an indirect methyltransferase inhibitor. Histone methyltransferases are a group of enzymes that catalyze the methylation of histone lysine and arginine by adding methyl groups to specific histone arginine or lysine residues. Other sizes are available. Please inqury us for quote.
Catalog No. NC1672427 Supplier Apexbio Technology LLC Supplier No. B612025
Adenosine dialdehyde (AdOx) has been identified as an indirect methyltransferase inhibitor. Histone methyltransferases are a group of enzymes that catalyze the methylation of histone lysine and arginine by adding methyl groups to specific histone arginine or lysine residues. Other sizes are available. Please inqury us for quote.