Learn More
Apexbio Technology LLC Adenosine Kinase Inhibitor (hydrate) 1mg
SDP
Catalog No. 5000089528
Encompass_Preferred
Adenosine Kinase Inhibitor (hydrate) is a small-molecule inhibitor targeting adenosine kinase It is designed to inhibit adenosine kinase thereby regulating intracellular adenosine metabolism Adenosine Kinase Inhibitor (hydrate) exerts its biological activity primarily through selective enzyme inhibition In cell-free conditions Adenosine Kinase Inhibitor (hydrate) demonstrates potent inhibitory activity with an IC50 value of approximately 1 7 nM Based on these pharmacological properties Adenosine Kinase Inhibitor (hydrate) holds research potential in investigating pain perception mechanisms and inflammation modulation through adenosine signaling pathways
Catalog No. 50-000-89528 Supplier Apexbio Technology LLC Supplier No. C52821
Adenosine Kinase Inhibitor (hydrate) is a small-molecule inhibitor targeting adenosine kinase It is designed to inhibit adenosine kinase thereby regulating intracellular adenosine metabolism Adenosine Kinase Inhibitor (hydrate) exerts its biological activity primarily through selective enzyme inhibition In cell-free conditions Adenosine Kinase Inhibitor (hydrate) demonstrates potent inhibitory activity with an IC50 value of approximately 1 7 nM Based on these pharmacological properties Adenosine Kinase Inhibitor (hydrate) holds research potential in investigating pain perception mechanisms and inflammation modulation through adenosine signaling pathways