Learn More
Medchemexpress LLC Adenosine monophosphate | 61-19-8 | 99.8% | C10H14N5O7P | 1 ML
SDP
Catalog No. 5000432423
Encompass_Preferred
Adenosine monophosphate (AMP) is an adenosine A1 receptor agonist and a key cellular metabolite involved in regulating energy homeostasis and signal transduction. It demonstrates significant antiviral activity against HSV-1 and HSV-2.
  • Acts as an adenosine A1 receptor agonist
  • Demonstrates significant antiviral activity against HSV-1 and HSV-2
  • Functions as a key cellular metabolite regulating energy homeostasis and signal transduction
  • Shows negligible cell toxicity at 25 to 400 μM in RAW264.7 cells
  • Significantly attenuates mRNA expression of TNF-α and IL-6 in RAW264.7 cells
  • Reduces recruitment of NF-κB p65 to TNF-α, IL-6, and IL-1β gene promoters
  • Increases hepatic adenosine levels in C57BL/6J mice
  • Improves survival rate and lowers serum AST and ALT levels in treated mice
  • Attenuates necrosis and reduces infiltration of inflammatory cells in vivo
Catalog No. 50-004-32423 Supplier Medchemexpress LLC Supplier No. HYA018110MM/1ML
Adenosine monophosphate (AMP) is an adenosine A1 receptor agonist and a key cellular metabolite involved in regulating energy homeostasis and signal transduction. It demonstrates significant antiviral activity against HSV-1 and HSV-2.
  • Acts as an adenosine A1 receptor agonist
  • Demonstrates significant antiviral activity against HSV-1 and HSV-2
  • Functions as a key cellular metabolite regulating energy homeostasis and signal transduction
  • Shows negligible cell toxicity at 25 to 400 μM in RAW264.7 cells
  • Significantly attenuates mRNA expression of TNF-α and IL-6 in RAW264.7 cells
  • Reduces recruitment of NF-κB p65 to TNF-α, IL-6, and IL-1β gene promoters
  • Increases hepatic adenosine levels in C57BL/6J mice
  • Improves survival rate and lowers serum AST and ALT levels in treated mice
  • Attenuates necrosis and reduces infiltration of inflammatory cells in vivo