Learn More
Medchemexpress LLC Adenosine monophosphate | 61-19-8 | MFCD00655082 | 97% | C10H14N5O7P | 10 G
SDP
Catalog No. 5000432247
Encompass_Preferred
Adenosine monophosphate is identified as an adenosine A1 receptor agonist. This compound exhibits significant antiviral activity against both HSV-1 and HSV-2. It is also recognized as a key cellular metabolite, playing a vital role in the regulation of energy homeostasis and signal transduction within biological systems.
  • Functions as an adenosine A1 receptor agonist.
  • Demonstrates significant antiviral activity against HSV-1 and HSV-2.
  • Acts as a key cellular metabolite.
  • Involved in regulating energy homeostasis.
  • Participates in signal transduction.
Catalog No. 50-004-32247 Supplier Medchemexpress LLC Supplier No. HYA018110G
Adenosine monophosphate is identified as an adenosine A1 receptor agonist. This compound exhibits significant antiviral activity against both HSV-1 and HSV-2. It is also recognized as a key cellular metabolite, playing a vital role in the regulation of energy homeostasis and signal transduction within biological systems.
  • Functions as an adenosine A1 receptor agonist.
  • Demonstrates significant antiviral activity against HSV-1 and HSV-2.
  • Acts as a key cellular metabolite.
  • Involved in regulating energy homeostasis.
  • Participates in signal transduction.