Learn More
Medchemexpress LLC Adenosine monophosphate | 61-19-8 | MFCD00655082 | 99.7% | C10H14N5O7P | 25 MG
SDP
Catalog No. 5000359163
Encompass_Preferred
Adenosine monophosphate is provided as an analytical standard, suitable for a variety of research and analytical applications. It functions as an adenosine A1 receptor agonist and exhibits significant antiviral activity against HSV-1 and HSV-2. As a key cellular metabolite, it plays a role in regulating energy homeostasis and signal transduction.
  • Intended for research and analytical applications
  • Functions as an adenosine A1 receptor agonist
  • Possesses significant antiviral activity against HSV-1 and HSV-2
  • Key cellular metabolite regulating energy homeostasis and signal transduction
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-003-59163 Supplier Medchemexpress LLC Supplier No. 25MGHYA0181R
Adenosine monophosphate is provided as an analytical standard, suitable for a variety of research and analytical applications. It functions as an adenosine A1 receptor agonist and exhibits significant antiviral activity against HSV-1 and HSV-2. As a key cellular metabolite, it plays a role in regulating energy homeostasis and signal transduction.
  • Intended for research and analytical applications
  • Functions as an adenosine A1 receptor agonist
  • Possesses significant antiviral activity against HSV-1 and HSV-2
  • Key cellular metabolite regulating energy homeostasis and signal transduction
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS