Learn More
Medchemexpress LLC Adenosine monophosphate (Standard) | 61-19-8 | 99.7% | C10H14N5O7P | 250 MG
SDP
Catalog No. 5000359180
Encompass_Preferred
Adenosine monophosphate (Standard) is the analytical standard for Adenosine monophosphate and is intended for research and analytical applications. It acts as an adenosine A1 receptor agonist, demonstrates significant antiviral activity against HSV-1 and HSV-2, and is a vital cellular metabolite involved in regulating energy homeostasis and signal transduction.
  • Analytical standard for research and analytical applications.
  • Functions as an adenosine A1 receptor agonist.
  • Exhibits significant antiviral activity against HSV-1 and HSV-2.
  • Key cellular metabolite for energy homeostasis and signal transduction.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-003-59180 Supplier Medchemexpress LLC Supplier No. 250MGHYA0181R
Adenosine monophosphate (Standard) is the analytical standard for Adenosine monophosphate and is intended for research and analytical applications. It acts as an adenosine A1 receptor agonist, demonstrates significant antiviral activity against HSV-1 and HSV-2, and is a vital cellular metabolite involved in regulating energy homeostasis and signal transduction.
  • Analytical standard for research and analytical applications.
  • Functions as an adenosine A1 receptor agonist.
  • Exhibits significant antiviral activity against HSV-1 and HSV-2.
  • Key cellular metabolite for energy homeostasis and signal transduction.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.