Learn More
Medchemexpress LLC Adenosine monophosphate (standard) | 61-19-8 | MFCD00655082 | 99.7% | C10H14N5O7P | 50 MG
SDP
Catalog No. 5000432481
Encompass_Preferred
Adenosine monophosphate (standard) is an analytical standard intended for research and analytical applications. It acts as an adenosine A1 receptor agonist, demonstrates significant antiviral activity against HSV-1 and HSV-2, and is a crucial cellular metabolite regulating energy homeostasis and signal transduction. This compound is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Acts as an adenosine A1 receptor agonist
  • Demonstrates significant antiviral activity against HSV-1 and HSV-2
  • Key cellular metabolite regulating energy homeostasis and signal transduction
  • Used as an analytical standard in qualitative research
  • Used as an analytical standard in quantitative research
  • Used as an analytical standard in methodological research experiments including HPLC, GC, and MS
Catalog No. 50-004-32481 Supplier Medchemexpress LLC Supplier No. HYA0181R50MG
Adenosine monophosphate (standard) is an analytical standard intended for research and analytical applications. It acts as an adenosine A1 receptor agonist, demonstrates significant antiviral activity against HSV-1 and HSV-2, and is a crucial cellular metabolite regulating energy homeostasis and signal transduction. This compound is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Acts as an adenosine A1 receptor agonist
  • Demonstrates significant antiviral activity against HSV-1 and HSV-2
  • Key cellular metabolite regulating energy homeostasis and signal transduction
  • Used as an analytical standard in qualitative research
  • Used as an analytical standard in quantitative research
  • Used as an analytical standard in methodological research experiments including HPLC, GC, and MS