Learn More
Medchemexpress LLC Adenosine-N,1,3,7,9-15N5 | 168566-57-2 | 99.9% | 1 MG
SDP
Catalog No. 5000406135
Encompass_Preferred
Adenosine-15N5 is a 15N labeled form of Adenosine, an endogenous autacoid that interacts with four G protein-coupled receptors (A1, A2A, A2B, and A3). This compound plays a role in various aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation. It is suitable for use as a tracer in drug development and for quantitative analysis.
  • Used as a tracer during drug development.
  • Serves as an internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, or LC-MS techniques.
  • Influences cellular processes like neuronal activity and vascular function.
  • Impacts platelet aggregation and blood cell regulation.
Catalog No. 50-004-06135 Supplier Medchemexpress LLC Supplier No. HYB0228S111MG
Adenosine-15N5 is a 15N labeled form of Adenosine, an endogenous autacoid that interacts with four G protein-coupled receptors (A1, A2A, A2B, and A3). This compound plays a role in various aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation. It is suitable for use as a tracer in drug development and for quantitative analysis.
  • Used as a tracer during drug development.
  • Serves as an internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, or LC-MS techniques.
  • Influences cellular processes like neuronal activity and vascular function.
  • Impacts platelet aggregation and blood cell regulation.