Learn More
Medchemexpress LLC Adenosine, N-benzoyl-, cyclic 3',5'-(hydrogen phosphate), sodium salt | 1135306-29-4 | 98.5% | 455.29 | C17H15N5NaO7P | 1 MG
SDP
Catalog No. 5000244867
Encompass_Preferred
6-Bnz-cAMP sodium salt is a cell-permeable cyclic AMP (cAMP) analog that selectively activates cAMP-dependent protein kinase A (PKA) without activating Epac-mediated signaling. It is supplied as the sodium salt solid for use in cell-based and biochemical assays to probe PKA-specific responses.
  • Cell-permeable cAMP analog for cellular and biochemical studies.
  • Selectively activates PKA but not Epac signaling pathways.
  • High purity (98.5%), suitable for sensitive assays.
  • Molecular weight approximately 455.29 g/mol.
  • Solid sodium salt form for enhanced solubility and handling.
  • Store sealed, away from moisture; recommended -20°C. In solvent: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-44867 Supplier Medchemexpress LLC Supplier No. HY1033221MG
6-Bnz-cAMP sodium salt is a cell-permeable cyclic AMP (cAMP) analog that selectively activates cAMP-dependent protein kinase A (PKA) without activating Epac-mediated signaling. It is supplied as the sodium salt solid for use in cell-based and biochemical assays to probe PKA-specific responses.
  • Cell-permeable cAMP analog for cellular and biochemical studies.
  • Selectively activates PKA but not Epac signaling pathways.
  • High purity (98.5%), suitable for sensitive assays.
  • Molecular weight approximately 455.29 g/mol.
  • Solid sodium salt form for enhanced solubility and handling.
  • Store sealed, away from moisture; recommended -20°C. In solvent: -80°C up to 6 months, -20°C up to 1 month.