Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine N1-oxide | 146-92-9 | MFCD00037993 | 99.5% | 283.24 g·mol⁻1 | C10H13N5O5 | 5 MG
SDP

Catalog No. 5000245346
Encompass_Preferred

Adenosine N1-oxide is a nucleoside derivative reported to have oral anti-inflammatory activity and is used in research on inflammation and cell differentiation. Supplied as a high-purity solid for laboratory use.

  • High purity (99.5%) solid, white to off-white.
  • Molecular weight 283.24 g·mol⁻1; formula C10H13N5O5.
  • Recommended storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Suitable for studies of osteogenic and adipocyte differentiation.
  • Available in milligram quantities for research applications.

Catalog No. 50-002-45346 Supplier Medchemexpress LLC Supplier No. HY1020825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine N1-oxide is a nucleoside derivative reported to have oral anti-inflammatory activity and is used in research on inflammation and cell differentiation. Supplied as a high-purity solid for laboratory use.

  • High purity (99.5%) solid, white to off-white.
  • Molecular weight 283.24 g·mol⁻1; formula C10H13N5O5.
  • Recommended storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Suitable for studies of osteogenic and adipocyte differentiation.
  • Available in milligram quantities for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.