Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine N1-oxide | 146-92-9 | MFCD00037993 | 99.5% | 283.24 g/mol | C10H13N5O5 | 10 MG
SDP

Catalog No. 5000245304
Encompass_Preferred

Adenosine N1-oxide is an adenosine derivative used in research with reported oral anti-inflammatory activity. Isolated from royal jelly, it has been reported to promote osteogenic and adipocyte differentiation and is supplied as a purified solid for laboratory studies.

  • Oral active anti-inflammatory agent.
  • Promotes osteogenic and adipocyte differentiation.
  • Molecular formula C10H13N5O5; molecular weight 283.24 g/mol.
  • Purity 99.5%.
  • Available in small milligram pack sizes for research use, e.g., 10 MG.
  • For research use only.

Catalog No. 50-002-45304 Supplier Medchemexpress LLC Supplier No. HY10208210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine N1-oxide is an adenosine derivative used in research with reported oral anti-inflammatory activity. Isolated from royal jelly, it has been reported to promote osteogenic and adipocyte differentiation and is supplied as a purified solid for laboratory studies.

  • Oral active anti-inflammatory agent.
  • Promotes osteogenic and adipocyte differentiation.
  • Molecular formula C10H13N5O5; molecular weight 283.24 g/mol.
  • Purity 99.5%.
  • Available in small milligram pack sizes for research use, e.g., 10 MG.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.