Learn More
TARGETMOL CHEMICALS INC Adenosine N1-oxide 5MG
SDP
Catalog No. 502877725
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Adenosine N1-oxide (1-Oxoadenosine), which is found in royal jelly, inhibited the secretion of inflammatory mediators by activated macrophages and reduced lethality in lipopolysaccharide (LPS)-induced endotoxin shock. Purity 99.24%
Catalog No. 50-287-7725 Supplier TARGETMOL CHEMICALS INC Supplier No. T236365MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Adenosine N1-oxide (1-Oxoadenosine), which is found in royal jelly, inhibited the secretion of inflammatory mediators by activated macrophages and reduced lethality in lipopolysaccharide (LPS)-induced endotoxin shock. Purity 99.24%