Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014764 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4764 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4764 Supplier MilliporeSigma Supelco Supplier No. PHR11381G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine is a purine nucleoside and a building block of RNA and many other biomolecules such as adenosine triphosphate and nicotinamide adenine dinucleotide. In the extracellular space, ecto-5′-nucleotidase (CD73) dephosphorylates adenosine triphosphate (ATP) to produce adenosine. Adenosine has four receptors namely A1R, A2AR A2BR and A3R. Adenosine plays a key role in the osteogenic differentiation. A1R induces osteoclast differentiation and A2AR induces osteoblast differentiation. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 58-61-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C10H13N5O4
MDL Number MFCD00005752
Melting Point 234°C to 236°C
Synonym 9-beta-D-Ribofuranosyladenine; Adenine riboside; Adenine-9-beta-D-ribofuranoside
Type Certified Reference Material
Quantity 1 g
Formula Weight 267.24
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.