Learn More
Medchemexpress LLC Adenosine receptor inhibitor 2 | 2550401-76-6 | 99.1% | 406.28 | 1 MG
SDP
Catalog No. 5000361328
Encompass_Preferred
This compound is a potent adenosine receptor (AR) inhibitor, demonstrating dual affinity for A1 and A2A ARs. It exhibits higher affinity for the A1AR compared to the A2AAR.
  • Potent AR inhibitor
  • Dual affinity for A1 and A2A ARs
  • Ki values: 52.2 nM for A1AR and 167 nM for A2AAR
  • For research use only
Catalog No. 50-003-61328 Supplier Medchemexpress LLC Supplier No. HY1479081MG
This compound is a potent adenosine receptor (AR) inhibitor, demonstrating dual affinity for A1 and A2A ARs. It exhibits higher affinity for the A1AR compared to the A2AAR.
  • Potent AR inhibitor
  • Dual affinity for A1 and A2A ARs
  • Ki values: 52.2 nM for A1AR and 167 nM for A2AAR
  • For research use only