Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine receptor inhibitor 2 | 2550401-76-6 | 99.1% | 406.28 | 25 MG
SDP

Catalog No. 5000363271
Encompass_Preferred

Adenosine receptor inhibitor 2 (compound 14b) is a potent AR (adenosine receptor) inhibitor that exhibits dual affinity toward A1/A2A ARs, with a higher affinity for A1AR than A2AAR. It has Ki values of 52.2 nM for A1AR and 167 nM for A2AAR.

  • Potent AR inhibitor.
  • Dual affinity for A1/A2A adenosine receptors.
  • Ki values of 52.2 nM for A1AR.
  • Ki values of 167 nM for A2AAR.
  • Targeted receptors include A1AR, A2AR, hA2B, and Adenosine A3 receptor.
  • Purity of 99.08%.
  • Molecular weight of 406.28.
  • Appearance as a white to off-white solid.

Catalog No. 50-003-63271 Supplier Medchemexpress LLC Supplier No. 25MGHY147908
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine receptor inhibitor 2 (compound 14b) is a potent AR (adenosine receptor) inhibitor that exhibits dual affinity toward A1/A2A ARs, with a higher affinity for A1AR than A2AAR. It has Ki values of 52.2 nM for A1AR and 167 nM for A2AAR.

  • Potent AR inhibitor.
  • Dual affinity for A1/A2A adenosine receptors.
  • Ki values of 52.2 nM for A1AR.
  • Ki values of 167 nM for A2AAR.
  • Targeted receptors include A1AR, A2AR, hA2B, and Adenosine A3 receptor.
  • Purity of 99.08%.
  • Molecular weight of 406.28.
  • Appearance as a white to off-white solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.