Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine receptor inhibitor 2 | 2550401-76-6 | 99.1% | 406.28 g/mol | C17H20BrN5O2 | 10 MG
SDP

Catalog No. 5000286090
Encompass_Preferred

Adenosine receptor inhibitor 2 is a small-molecule research compound that acts as an adenosine receptor inhibitor with reported dual affinity for A1 and A2A receptors. It is supplied as a high-purity analytical standard intended for pharmacology and receptor-binding studies.

  • High-purity small-molecule suitable for receptor pharmacology.
  • Dual affinity for A1 and A2A adenosine receptors.
  • Chemical formula C17H20BrN5O2, molecular weight 406.28 g/mol.
  • CAS number 2550401-76-6 for substance identification.
  • Available in milligram-scale packages, including 10 mg.
  • Structural identifiers available (SMILES) for cheminformatics use.
  • For research use only; not for human consumption or clinical use.

Catalog No. 50-002-86090 Supplier Medchemexpress LLC Supplier No. HY14790810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine receptor inhibitor 2 is a small-molecule research compound that acts as an adenosine receptor inhibitor with reported dual affinity for A1 and A2A receptors. It is supplied as a high-purity analytical standard intended for pharmacology and receptor-binding studies.

  • High-purity small-molecule suitable for receptor pharmacology.
  • Dual affinity for A1 and A2A adenosine receptors.
  • Chemical formula C17H20BrN5O2, molecular weight 406.28 g/mol.
  • CAS number 2550401-76-6 for substance identification.
  • Available in milligram-scale packages, including 10 mg.
  • Structural identifiers available (SMILES) for cheminformatics use.
  • For research use only; not for human consumption or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.