Learn More
Medchemexpress LLC Adenosine (Standard) | 58-61-7 | 99.39% | 50 MG
SDP
Catalog No. 5000405655
Encompass_Preferred
Adenosine (Standard) is the analytical standard of Adenosine, intended for research and analytical applications. Adenosine (Adenine riboside), a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors: A1, A2A, A2B, and A3. It affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.
  • Analytical standard for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-004-05655 Supplier Medchemexpress LLC Supplier No. HYB0228R50MG
Adenosine (Standard) is the analytical standard of Adenosine, intended for research and analytical applications. Adenosine (Adenine riboside), a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors: A1, A2A, A2B, and A3. It affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.
  • Analytical standard for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS