Learn More
Medchemexpress LLC Adenosine (standard) | 58-61-7 | >99.4% | 100 MG
SDP
Catalog No. 5000405743
Encompass_Preferred
Adenosine (Standard) is the analytical standard of Adenosine, intended for research and analytical applications. Adenosine (Adenine riboside) is a ubiquitous endogenous autacoid that influences nearly all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.
  • Analytical standard for research and analytical applications
  • Acts through G protein-coupled receptors A1, A2A, A2B, and A3
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-004-05743 Supplier Medchemexpress LLC Supplier No. HYB0228R100MG
Adenosine (Standard) is the analytical standard of Adenosine, intended for research and analytical applications. Adenosine (Adenine riboside) is a ubiquitous endogenous autacoid that influences nearly all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.
  • Analytical standard for research and analytical applications
  • Acts through G protein-coupled receptors A1, A2A, A2B, and A3
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS