Learn More
Medchemexpress LLC Adenosine (standard) | 58-61-7 | 99.4% | 25 MG
SDP
Catalog No. 5000450643
Encompass_Preferred
Adenosine (Standard) is the analytical standard of Adenosine, intended for research and analytical applications. Adenosine (Adenine riboside) is a ubiquitous endogenous autacoid that acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. It affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.
  • Analytical standard of Adenosine
  • Intended for research and analytical applications
  • Acts through four G protein-coupled receptors: A1, A2A, A2B, and A3
  • Affects neuronal activity, vascular function, platelet aggregation, and blood cell regulation
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-50643 Supplier Medchemexpress LLC Supplier No. 25MGHYB0228R
Adenosine (Standard) is the analytical standard of Adenosine, intended for research and analytical applications. Adenosine (Adenine riboside) is a ubiquitous endogenous autacoid that acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. It affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.
  • Analytical standard of Adenosine
  • Intended for research and analytical applications
  • Acts through four G protein-coupled receptors: A1, A2A, A2B, and A3
  • Affects neuronal activity, vascular function, platelet aggregation, and blood cell regulation
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS