Learn More
Medchemexpress LLC Adenosylhomocysteinase (SAHH; AHCY) | 9025-54-1 | ≥140 U/mg protein | 44 kDa | 5 MG
SDP
Catalog No. 5000361122
Encompass_Preferred
Adenosylhomocysteinase (SAHH; AHCY) is a highly conserved enzyme that reversibly catalyzes S-adenosylhomocysteine (SAH) to adenosine and L-homocysteine. It has a molecular weight of 44 kDa, is stable in the pH range of 6.0-8.0, and is intended for research use only.
  • Reversibly catalyzes S-adenosylhomocysteine (SAH) to adenosine and L-homocysteine
  • Serum exosomal level acts as a prognostic biomarker in HBV-LC patients
  • Obtained by microbial recombination
  • Stable in pH range of 6.0-8.0
  • Reconstitute with pure water containing 20% glycerol
  • Store at 4°C for up to one year after reconstitution
  • For extended storage, aliquot and freeze at -20°C; avoid repeated freeze-thaw cycles
Catalog No. 50-003-61122 Supplier Medchemexpress LLC Supplier No. HY1490485MG
Adenosylhomocysteinase (SAHH; AHCY) is a highly conserved enzyme that reversibly catalyzes S-adenosylhomocysteine (SAH) to adenosine and L-homocysteine. It has a molecular weight of 44 kDa, is stable in the pH range of 6.0-8.0, and is intended for research use only.
  • Reversibly catalyzes S-adenosylhomocysteine (SAH) to adenosine and L-homocysteine
  • Serum exosomal level acts as a prognostic biomarker in HBV-LC patients
  • Obtained by microbial recombination
  • Stable in pH range of 6.0-8.0
  • Reconstitute with pure water containing 20% glycerol
  • Store at 4°C for up to one year after reconstitution
  • For extended storage, aliquot and freeze at -20°C; avoid repeated freeze-thaw cycles