Learn More
Enzo Life Sciences Adenovirus type 5 hexon protein (100µg)
SDP
Supplier:  Enzo Life Sciences ALX6301200100
Encompass_Preferred
Native protein. Native protein. Purity: ≥95% (SDS-PAGE). Appearance: Clear liquid. Formulation: Liquid. In 10mM BIS-TRIS propane, containing 375mM sodium chloride and 0.09% sodium azide. Source: From adenovirus type 5. Long Term Storage: -80°C. Applications: ELISA Positive control, Antibody capture.
Catalog No. 50-200-9647