Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenovirus 9 E4-Orf1 Synthetic Peptide

Catalog No. PEP0721 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0721 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0721 Supplier Invitrogen™ Supplier No. PEP0721
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 16 amino acids near the carboxy terminus of Adenovirus-9 E4 Orf1.

Adenoviruses are DNA viruses generally widespread in nature that are frequently the cause of acute upper respiratory tract infections (i.e. common colds). 49 human adenovirus serotypes have been discovered and divided into six subgroups, A to E, based on observation. They are an important cause of respiratory illness, conjunctivitis, gastroenteritis, and have more recently been recognized as a potential cause of sexually transmitted diseases. Several types have oncogenic potential though most cause self-limiting febrile illnesses characterized by inflammation of conjunctivae and the respiratory tract. The virus can be isolated from the majority of tonsils/adenoids surgically removed, indicating latent infections. It is not known how long the virus can persist in the body, or whether it is capable of reactivation after long periods. In patients experiencing immunosuppression (e.g. AIDS) it can be reactivated, causing disease. Adenovirus vectors, particularly those based on Ad5, have shown great promise for the development of gene therapy protocols and have been used for clinical purpose.
TRUSTED_SUSTAINABILITY

Specifications

Species Virus
Purification Method purified
Common Name Adenovirus 9 E4-Orf1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias Adeno Virus; Adenovirus Type 9; AdV; HADV
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.