Learn More
Drg International Inc DRG Adenovirus IgA ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3445R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgA-class antibodies to Adenovirus in serum and heparin plasma. This product is for Research Use Only. The DRG Adenovirus IgA ELISA kit is a solid phase enzyme linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with inactivated Adenovirus Grade 2 antigen (strain Adenoid 6). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyper-immune anti-human IgG-class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors.
Catalog No. 50-194-7554