Learn More
Drg International Inc Adenovirus Igg Elisa
Supplier:  Drg International Inc EIA3446
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Adenovirus in serum. This assay is intended for in vitro diagnostic use only. Adenoviruses are double-stranded DNA viruses lacking an envelope, of about 60-90 nm. Adenoviruses were first isolated from adenoid tissue and have a certain affinity for lymph glands, where they may remain latent for years. They also invade the respiratory tract, the gastrointestinal tract, and the conjunctiva. Adenovirus infections are widely distributed and common with most infections occurring in childhood. The DRG Adenovirus IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with inactivated Adenovirus Grade 2 antigen (strain Adenoid 6). Diluted patient specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7555