Learn More
Drg International Inc DRG Adenovirus IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3447R
Encompass_Preferred
An enzyme immunoassay for the qualitative determination of IgM-class antibodies to Adenovirus in human serum and heparin plasma. This product is for Research Use Only. The DRG Adenovirus IgM ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyperimmune anti-human IgG-class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors.
Catalog No. 50-194-7556