Learn More
Enzo Life Sciences Adenovirus type 40 antigen (100µg)
SDP
Supplier:  Enzo Life Sciences ALX6301210100
Encompass_Preferred
Native protein. Native protein. Purity: ≥99%. Appearance: Clear liquid. Formulation: Liquid. In PBS. Source: From adenovirus type 40. Inactivated by 2 hours at 54°C. Long Term Storage: -80°C. Applications: ELISA LINE assay, Immunogen, Antibody capture.
Catalog No. 50-200-9648