Learn More
Cayman Chemical Adenylate Cyclase ToxIn50ug
SDP
Catalog No. 502938410
Encompass_Preferred
A virulence factor in B. pertussis; has been used in the development of a microsphere-based multiplex antibody capture assay to quantify antibody amounts in acellular pertussis vaccines
Catalog No. 50-293-8410 Supplier Cayman Chemical Supplier No. 2821150 UG
Add to Cart
Add to Cart
A virulence factor in B. pertussis; has been used in the development of a microsphere-based multiplex antibody capture assay to quantify antibody amounts in acellular pertussis vaccines