Learn More
Cayman Chemical Adenylosuccinic Acid, Purity: ≥95%, CAS: 19046-78-7, 50 mg
SDP
Catalog No. 502989230
Encompass_Preferred
A purine nucleotide and an intermediate in the purine nucleotide cycle; converted into AMP and fumarate by ADSL in the cytosol; inhibits calcium-induced activation of non-selective cation channels in isolated rat brown adipocytes in a patch-clamp assay at 10 µM; induces contractions in isolated guinea pig uterus strips at 100 µM; increases glucose-induced insulin exocytosis in INS-1 832/13 insulinoma cells at 10 µM; decreases muscle damage and pseudohypertrophy in a dystrophin-deficient mdx mouse model of DMD
Catalog No. 50-298-9230 Supplier Cayman Chemical Supplier No. 2969650 MG
A purine nucleotide and an intermediate in the purine nucleotide cycle; converted into AMP and fumarate by ADSL in the cytosol; inhibits calcium-induced activation of non-selective cation channels in isolated rat brown adipocytes in a patch-clamp assay at 10 µM; induces contractions in isolated guinea pig uterus strips at 100 µM; increases glucose-induced insulin exocytosis in INS-1 832/13 insulinoma cells at 10 µM; decreases muscle damage and pseudohypertrophy in a dystrophin-deficient mdx mouse model of DMD