Learn More
Medchemexpress LLC Adenylosuccinic acid tetraammonium | >98% | 10 MG
SDP
Catalog No. 5000342778
Encompass_Preferred
Adenylosuccinic acid tetraammonium is an orally active purine ribonucleoside monophosphate involved in nucleotide cycle metabolism. This compound can be converted into fumaric acid via adenylosuccinate lyase and holds potential for the study of Duchenne muscular dystrophy (DMD).
  • Orally active purine ribonucleoside monophosphate
  • Plays a role in nucleotide cycle metabolism
  • Converts into fumaric acid via adenylosuccinate lyase
  • Potential for Duchenne muscular dystrophy (DMD) research
  • Ameliorates murine Duchenne muscular dystrophy symptoms in vivo
Catalog No. 50-003-42778 Supplier Medchemexpress LLC Supplier No. HY127137A10MG
Adenylosuccinic acid tetraammonium is an orally active purine ribonucleoside monophosphate involved in nucleotide cycle metabolism. This compound can be converted into fumaric acid via adenylosuccinate lyase and holds potential for the study of Duchenne muscular dystrophy (DMD).
  • Orally active purine ribonucleoside monophosphate
  • Plays a role in nucleotide cycle metabolism
  • Converts into fumaric acid via adenylosuccinate lyase
  • Potential for Duchenne muscular dystrophy (DMD) research
  • Ameliorates murine Duchenne muscular dystrophy symptoms in vivo