Learn More
Medchemexpress LLC Adenylyl-imidodiphosphate disodium | 7414-56-4 | MFCD32646673 | 99.4% | 549.17 | C11H16N5Na2O12P3 | 10 MG
SDP
Catalog No. 5000240757
Encompass_Preferred
AMP-PCP disodium is a non-hydrolyzable ATP analogue (adenylyl-imidodiphosphate disodium) used to mimic ATP binding in biochemical and structural studies. It binds the N-terminal domain of Hsp90 with a reported Kd of 3.8 μM and favors formation of the active Hsp90 homodimer, making it useful for probing ATP-dependent enzyme mechanisms and protein complex assembly.
  • Non-hydrolyzable ATP analogue for stabilizing ATP-bound states.
  • Binds Hsp90 N-terminal domain (Kd ≈ 3.8 μM), facilitating homodimer formation.
  • Provided as the disodium salt with high purity (99.4%).
  • High aqueous solubility: H2O 100 mg/mL; PBS 50 mg/mL (ultrasonic recommended).
  • Storage recommendations: solid -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Supplied in small research pack sizes suitable for biochemical assays.
Catalog No. 50-002-40757 Supplier Medchemexpress LLC Supplier No. HY106723A10MG
AMP-PCP disodium is a non-hydrolyzable ATP analogue (adenylyl-imidodiphosphate disodium) used to mimic ATP binding in biochemical and structural studies. It binds the N-terminal domain of Hsp90 with a reported Kd of 3.8 μM and favors formation of the active Hsp90 homodimer, making it useful for probing ATP-dependent enzyme mechanisms and protein complex assembly.
  • Non-hydrolyzable ATP analogue for stabilizing ATP-bound states.
  • Binds Hsp90 N-terminal domain (Kd ≈ 3.8 μM), facilitating homodimer formation.
  • Provided as the disodium salt with high purity (99.4%).
  • High aqueous solubility: H2O 100 mg/mL; PBS 50 mg/mL (ultrasonic recommended).
  • Storage recommendations: solid -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Supplied in small research pack sizes suitable for biochemical assays.