Learn More
Medchemexpress LLC Adenylyl-imidodiphosphate tetralithium | 72957-42-7 | ≥97.0% | 529.93 g·mol⁻¹ | C10H13Li4N6O12P3 | 5 MG
SDP
Catalog No. 5000262453
Encompass_Preferred
AMP-PNP tetralithium (adenylyl-imidodiphosphate tetralithium) is a non-hydrolyzable ATP analog supplied as the tetralithium salt. It binds to ATP-binding sites without undergoing enzymatic hydrolysis, enabling stabilization of nucleotide-bound states for biochemical assays, structural studies, and investigations of ATP-dependent processes.
  • Non-hydrolyzable ATP analog for probing ATP-dependent processes.
  • Stabilizes nucleotide-bound protein conformations for structural studies.
  • Suitable for enzyme assays, kinase regulation, and ion-channel research.
  • Tetralithium salt form for use in appropriate buffer systems.
  • High purity (≥97.0%) appropriate for research applications.
  • Molecular weight 529.93 g·mol⁻¹; formula C10H13Li4N6O12P3.
Catalog No. 50-002-62453 Supplier Medchemexpress LLC Supplier No. HY1289335MG
AMP-PNP tetralithium (adenylyl-imidodiphosphate tetralithium) is a non-hydrolyzable ATP analog supplied as the tetralithium salt. It binds to ATP-binding sites without undergoing enzymatic hydrolysis, enabling stabilization of nucleotide-bound states for biochemical assays, structural studies, and investigations of ATP-dependent processes.
  • Non-hydrolyzable ATP analog for probing ATP-dependent processes.
  • Stabilizes nucleotide-bound protein conformations for structural studies.
  • Suitable for enzyme assays, kinase regulation, and ion-channel research.
  • Tetralithium salt form for use in appropriate buffer systems.
  • High purity (≥97.0%) appropriate for research applications.
  • Molecular weight 529.93 g·mol⁻¹; formula C10H13Li4N6O12P3.