Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX Diagnostics ADEXUSDx hCG Control Set

Catalog No. 21381503
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-381-503 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-381-503 Supplier MEDTOX Diagnostics Supplier No. 600735
Add to Cart
Add to Cart
This item is not returnable. View return policy

Control Set is an unassayed quality control (QC) set for monitoring the reliability of tests for the detection of hCG.

ADEXUSDx hCG Control Set is an unassayed quality control (QC) set for monitoring the reliability of tests for the detection of hCG.

Specifications

Sample Type Whole Blood, Serum, Plasma
Type hCG Control Set
Disposable Single-use
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.