Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOW Diagnostics ADEXUSDx™ hCG Test
SDP

Catalog No. 23049002
Encompass
Change view
Click to view available options
Quantity:
1 box - 25 tests in a box
1 box - 50 tests in a box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-049-002 1 box - 50 tests in a box
23-049-000 1 box - 25 tests in a box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-049-002 Supplier NOW Diagnostics Supplier No. 800150

missing translation for 'validMainframeMsg'

The ADEXUSDx™ hCG Test is an immunoassay used for the qualitative detection of human chorionic gonadotropin (hCG) in human whole blood, plasma, or serum that delivers results in ten minutes without additional reagents or materials.

  • Uses whole blood (capillary or venous), plasma, or serum
  • >99% sensitivity and specificity
  • Clinical relevant cutoff: 10 mIU/mL
  • Small sample volume test (about 40 μL)
  • Results in 10 minutes, no waiting on urine samples
  • No reagents or buffers
  • Reduced exposure to liability from false negatives
  • Increased patient throughput 

Certifications

FDA Approved in USA; CE Marked in Europe

Specifications

Content And Storage Stable unopened at 4°C-30°C (39°F-86°F) until the expiration date
CE Marker Yes
Clia Complexity Moderate (Waiver Pending)
Detectable Analytes hCG
Sample Type Capillary or Venous blood/whole blood/plasma/serum
Sensitivity 10 mIU/mL
Type Rapid Diagnostic Lateral Flow Assay
Disposable Single-use
Formulation The device contains material of human or animal origin
For Use With (Application) For in vitro diagnostic use only
Includes Product Insert
Packaging Type Packaged in individually sealed pouches
Quantity 1 box - 50 tests in a box
Range Clinical relevant cutoff: 10 mIU/mL
Test Time 10 min.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.