Learn More
Selleck Chemical LLC Adezmapimod hydrochloride E4791-1G
SDP
Catalog No. 5000039472
Encompass_Preferred
Adezmapimod hydrochloride (RWJ 64809 hydrochloride SB 203580 hydrochloride) is an inhibitor of the MAPK family member termed as stress-activated protein kinase 2a(SAPK2a also known as p38) It exhibits IC50 of SAPK2a/p38 and SAPK2b/p38 2 with IC50 values of 50 nM and 500 nM respectively and can be used in research of chronic inflammatory diseases such as rheumatoid arthritis
Catalog No. 50-000-39472 Supplier Selleck Chemical LLC Supplier No. E47911G
Adezmapimod hydrochloride (RWJ 64809 hydrochloride SB 203580 hydrochloride) is an inhibitor of the MAPK family member termed as stress-activated protein kinase 2a(SAPK2a also known as p38) It exhibits IC50 of SAPK2a/p38 and SAPK2b/p38 2 with IC50 values of 50 nM and 500 nM respectively and can be used in research of chronic inflammatory diseases such as rheumatoid arthritis