Learn More
3M™ Adflo™ Replacement Cartridges 15-0499-99X06, Organic Vapor/Acid Gas
Catalog No. 19153731 Shop All 3M Products
Change view
Click to view available options
6/Case
Catalog No.
19-153-731 6/Case
1 options
Catalog No. 19-153-731 Supplier 3M™ Supplier No. 7000128119
Add to Cart
Add to Cart
For use with 3M Adflo Powered Air Purifying Respirator (PAPR).
  • Cartridge can be stacked onto the high efficiency particulate filter for additional protection against:
    • Organic vapors
    • Sulfur dioxide
    • Chlorine
    • Hydrogen chloride
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cartridge
For Use With (Equipment) 3M Adflo Powered Air Purifying Respirator
Model 15-0499-99X06
Quantity 6/Cs.