Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Adflo™ Powered Air Purifying Respirator Suspenders

Catalog No. 19072166 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-166 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-166 Supplier 3M™ Supplier No. 7100215094
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PAPR replacement suspenders for use if the original suspenders becomes worn out or damaged.

  • For maximum safety and comfort
  • Check your equipment regularly
  • Replace worn out or damaged parts
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Disposable Reusable
Product Type Suspenders
For Use With (Application) Grinding, Welding, Metal Repair, Metal Processing
For Use With (Equipment) G5-01 Welding Helmets
Model 15-0099-17
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.