Learn More
Cayman Chemical aDGalactopyrnosylphenyl Is
SDP
Catalog No. 502984242
Encompass_Preferred
Used to prepare various neoglycoproteins, which consist of a glycosylated serum albumin substituted with either fluorescein or methotrexate
Catalog No. 50-298-4242 Supplier Cayman Chemical Supplier No. 19544500 UG
Used to prepare various neoglycoproteins, which consist of a glycosylated serum albumin substituted with either fluorescein or methotrexate