Learn More
Cayman Chemical α-D-Galactose-1-phosphate (potassium salt hydrate), Purity: ≥95%, 100 mg
SDP
Catalog No. 502967248
Encompass_Preferred
An intermediate in the interconversion of glucose and galactose
Catalog No. 50-296-7248 Supplier Cayman Chemical Supplier No. 17863100 MG
An intermediate in the interconversion of glucose and galactose