Learn More
Cayman Chemical aDGlucose1 6bIsphosphate 10mg
SDP
Catalog No. 502970455
Encompass_Preferred
A bis-phosphorylated derivative of α-D-glucose; has roles in carbohydrate metabolism; the product of the reaction of glucose 1- or 6-phosphate with PGM2LI in the conversion of 1,3-bisphosphoglycerate to 3-phosphoglycerate; a cofactor for the bacterial enzyme phosphopentomutase; has been used in the study of carbohydrate metabolism
Catalog No. 50-297-0455 Supplier Cayman Chemical Supplier No. 1646410 MG
A bis-phosphorylated derivative of α-D-glucose; has roles in carbohydrate metabolism; the product of the reaction of glucose 1- or 6-phosphate with PGM2LI in the conversion of 1,3-bisphosphoglycerate to 3-phosphoglycerate; a cofactor for the bacterial enzyme phosphopentomutase; has been used in the study of carbohydrate metabolism