Learn More
Apexbio Technology LLC ADH-503 2055362-74-6 1mL(10 mM in DMSO)
SDP
Catalog No. 5000088028
Encompass_Preferred
ADH-503 (CAS 2055362-74-6) also known as (Z)-Leukadherin-1 choline is an orally bioavailable small molecule that functions as an allosteric agonist This compound modulates macrophage phenotypes by promoting repolarization of tumor-associated macrophages resulting in a reduction of immunosuppressive myeloid cell infiltration within the tumor microenvironment Additionally ADH-503 has been shown to enhance dendritic cell responses These properties make ADH-503 a valuable tool for investigating macrophage regulation and immune modulation in the context of cancer research
Catalog No. 50-000-88028 Supplier Apexbio Technology LLC Supplier No. BA58845.1
ADH-503 (CAS 2055362-74-6) also known as (Z)-Leukadherin-1 choline is an orally bioavailable small molecule that functions as an allosteric agonist This compound modulates macrophage phenotypes by promoting repolarization of tumor-associated macrophages resulting in a reduction of immunosuppressive myeloid cell infiltration within the tumor microenvironment Additionally ADH-503 has been shown to enhance dendritic cell responses These properties make ADH-503 a valuable tool for investigating macrophage regulation and immune modulation in the context of cancer research