Learn More
TARGETMOL CHEMICALS INC ADH-503 25MG
SDP
Catalog No. 502576286
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ADH-503 (GB1275) is a potent agonist of the integrin CD11b to mitigate myeloid cell immunosuppression. purity: 98%
Catalog No. 50-257-6286 Supplier TARGETMOL CHEMICALS INC Supplier No. T777625MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ADH-503 (GB1275) is a potent agonist of the integrin CD11b to mitigate myeloid cell immunosuppression. purity: 98%