Learn More
Medchemexpress LLC HY-15701B 5mg Medchemexpress, ADH-503 CAS:2055362-74-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15701B5MG
Encompass_Preferred
Medchemexpress, HY-15701B 5mg ADH-503 CAS:2055362-74-6 ADH-503 ((Z)-Leukadherin-1 choline) is an orally active and allosteric CD11b agonist. ADH-503 leads to the repolarization of tumor-associated macrophages, reduction in the number of tumor-infiltrating immunosuppressive myeloid cells, and enhances dendritic cell responses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0671