Learn More
Aviva Systems Biology Corp ADH1A Antibody - N-terminal region (ARP41785_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41785_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADH1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ADH1A. Alternate names: ADH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57992