Learn More
Aviva Systems Biology Corp ADH1B Antibody - N-terminal region (ARP41786_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41786_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADH1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ADH1B. Alternate names: ADH2, DKFZp686C06125ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57993