Learn More
Aviva Systems Biology Corp ADH4 Antibody - middle region (ARP43573_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43573_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ADH4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ADH4. Alternate names: ADH-2ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Pig, Rabbit, Rat
Catalog No. 50-014-58574