Learn More
Aviva Systems Biology Corp ADH4 Antibody - middle region (ARP43572_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43572_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADH4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ADH4. Alternate names: ADH-2ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-58573